Learn More
Cytiva HyClone™ Phosphate Buffered Saline (PBS)
Product Code. 10462372
Change view
Click to view available options
Quantity:
500 mL
1000 mL
6 x 500 mL
6 x 1000 mL
Concentration:
1 X
10 X
Unit Size:
1000mL
500mL
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Concentration Unit Size
10462372 500 mL 1 X 500mL
10572193 1000 mL 1 X 1000mL
10793004 6 x 1000 mL 1 X Pack of 6
10294922 500 mL 10 X 500mL
10356662 1000 mL 10 X 1000mL
10126473 6 x 500 mL 1 X Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10462372 Supplier Cytiva Supplier No. SH30256.01
  • ONLINE EXCLUSIVE PRICE
    30.24 EUR
    25.00 EUR / 500mL
    Save 5.24 EUR
    17% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Buffer cell culture growth with PBS
  • QC-tested solution
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Content And Storage 15° to 30°C
Concentration 1 X
Form Liquid
Product Type Phosphate Buffered Saline (PBS)
Sterilization Method 0.1 μm Sterile-filtered
Without Additives No Calcium, No Magnesium, No Phenol Red
Endotoxin Level =1.0EU/mL
pH 7.0 to 7.2
Quantity 500 mL