Learn More
Cytiva HyClone™ Fetal Bovine Serum, South American Origin
Brand:  Cytiva SV30160.03
314.47 EUR valid until 2024-12-31
Use promo code "23281" to get your promotional price.
Additional Details : Weight : 0.54000kg
Product Code. 10309433
  • 338.00 EUR / 500mL
Estimated Shipment: 22-05-2024
Description
  • Filtered through three sequential 100nm (0.1μm) pore-size rated filters
  • Not sold in the U. S. Only for sale to European and Asian customers