Learn More
Cytiva HyClone™ Donor Equine Serum, US Origin, Irradiated 30-45 kGy
Product Code. 17642075
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17642075 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17642075 Supplier Cytiva Supplier No. SH30074.04IR3045
  • ONLINE EXCLUSIVE PRICE
    179.00 EUR
    132.63 EUR / 1 L
    Save 46.37 EUR
    25% Off
 Restricted Item
Estimated Shipment: 03-02-2027
Complete traceability back to the original source.
  • Tested for equine infectious anemia
  • Complete traceability back to original source
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C.
Country of Origin United States
Quantity 1000 mL
Sterilization Method Irradiated
Endotoxin Concentration ≤ 10 EU/mL
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Not Detected