Learn More
Cytiva HyClone™ Donor Equine Serum, US Origin, Irradiated 25-40 kGy, Heat-Inactivated
Product Code. 17632075
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17632075 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17632075 Supplier Cytiva Supplier No. SH30074.03IH2540
  • ONLINE EXCLUSIVE PRICE
    135.00 EUR
    99.84 EUR / 500mL
    Save 35.16 EUR
    26% Off
 Restricted Item
Estimated Shipment: 23-02-2027
Complete traceability back to the original source.
  • Tested for equine infectious anemia
  • Complete traceability back to original source
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C.
Country of Origin United States
Quantity 500 mL
Sterilization Method Irradiated
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Not Detected