Learn More
Cytiva HyClone™ Cosmic Calf Serum (U.S. origin)
Product Code. 12694227
Change view
Click to view available options
Quantity:
100 mL
1000 mL
Unit Size:
1000mL
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12694227 1000 mL 1000mL
12349842 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12694227 Supplier Cytiva Supplier No. SH30087.04HI
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Currently Unavailable: We have no information when this product will be available.
A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.
HyClone Cosmic Calf serum is a high-performing bovine calf serum that has been fortified with iron and a proprietary blend of growth-promoting factors. The result is a naturally derived, high-quality engineered serum that has demonstrated excellent performance with a broad range of cell types. All components of Cosmic Calf serum are sourced in the USA and are traceable.
TRUSTED_SUSTAINABILITY
Spezifikation
Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Color Amber
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
Species Bovine
Age 16 to 22 Weeks
Country of Origin United States
Form Frozen
Format Bottle
Additive Iron, Naturally Derived Components
Product Type Calf Bovine Serum
Quantity 1000 mL
Serum Treatment Heat-inactivated
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm, Gamma-irradiated
Processing Heat Inactivated
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration <30 mg/dL
Iron Supplemented Yes
Osmolality 280 to 360 mOsm/kg
USDA Safety Testing Included No
Gamma-irradiated No
Mehr anzeigen Weniger anzeigen