Learn More
Cytiva HyClone™ CDM4NS0 Cell Culture Media
Product Code. 10153202
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10153202 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10153202 Supplier Cytiva Supplier No. SH30578.01
  • ONLINE EXCLUSIVE PRICE
    475.00 EUR
    366.56 EUR / 5 L
    Save 108.44 EUR
    22% Off
 Restricted Item
In Stock
Regulatory-friendly medium designed to increase process yields with a variety of NS0 cell clones.
  • Hydrolysate-free and animal-derived component-free formulation.
  • Developed for high cell yield and MAb production.
  • Ready-to-use formulation, requires no cholesterol or GS supplementation.
  • Allows for direct or sequential adaptation.
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies.
  • Manufactured from traceable components according to cGMP guidelines.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Form Powder
    Sterility Non-sterile
    Without Additives No Glutamine, No Hydrolysate, No Phenol Red, No Protein, No HEPES, No Sodium Bicarbonate
    Quantity 5 L
    Format 5 L bottle
    Content And Storage 2° to 8°C, away from light and moisture