Learn More
Cytiva HyClone™ Bovine Calf Serum (BCS), US Origin, Irradiated 25-40 kGy
Product Code. 17622075
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17622075 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17622075 Supplier Cytiva Supplier No. SH30073.04IR2540
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
A cost-effective alternative to Fetal Bovine Serum (FBS).
  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Age of animal at time of collection approximately 16-24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C.
Country of Origin United States
Quantity 1000 mL
Sterilization Method Irradiated
Endotoxin Concentration ≤ 10 EU/mL
Hemoglobin Concentration < 30 mg/dL
Mycoplasma Detected Not Detected