Learn More
Cytiva HyClone™ Amphotericin B (Fungiezone) Solution
Product Code. 11526481
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11526481 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11526481 Supplier Cytiva Supplier No. SV30078.01
  • ONLINE EXCLUSIVE PRICE
    45.92 EUR
    42.77 EUR / 50mL
    Save 3.15 EUR
    6% Off
 Restricted Item
In Stock
  • An antibiotic agent that is active against fungi
  • Used as an antifungal, anti-yeast and anti-mold agent
  • Interferes with the permeability of the cell membrane of sensitive fungi by binding sterols
  • Common working concentration is 2.5μg/mL, and it is cytotoxic at 30μg/mL
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store frozen
Description Amphotericin B (Fungizone) Solution, 250μg/mL
Antibiotic Spectrum Antimycotic, Antifungal
Concentration 250 μg/mL
Form Liquid
Product Type Antifungal
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 50 mL