Learn More
Stuart™ Hybridization Bottle
Product Code. 11456558
Change view
Click to view available options
Length (Metric):
300 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11456558 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11456558 Supplier Stuart™ Supplier No. 1623799
  • ONLINE EXCLUSIVE PRICE
    201.00 EUR
    177.67 EUR / Pack of 1
    Save 23.33 EUR
    11% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Stuart™ Hybridization Bottle for use with Stuart Hybridization Incubators, and Shakers
  • Choose from a wide range of accessories for Stuart Hybridization Incubators
  • Different capacity rotisseries and bottles are available to suit specific applications
Specifications
Color Clear, with red cap
Diameter (Metric) Inner 40 mm
Length (Metric) 300 mm
Material Glass, Polyester, PTFE
Product Type Hybridization Bottle