Learn More
Cytiva Hybond™ Blotting Paper
Product Code. 11750094 Shop All Cytiva Products
Change view
Click to view available options
Type:
Hybond Blotting Paper
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11750094 Hybond Blotting Paper Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11750094 Supplier Cytiva Supplier No. RPN6101M
  • ONLINE EXCLUSIVE PRICE
    102.83 EUR
    89.17 EUR / Pack of 100
    Save 13.66 EUR
    13% Off
In Stock
Use with Hybond™ blotting membranes. Cytiva Life Sciences™ Hybond™ Blotting Paper is pure cellulose grade and perfect for gel electrophoresis applications.
  • For gel electrophoresis
  • Used with Hybond blotting membranes
  • Pure cellulose grade
  • Medium wet strength
  • Smooth surface
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Gel Electrophoresis, Hybond Blotting Membranes
Type Hybond Blotting Paper
Thickness (English) 0.01 in.
Thickness (Metric) 0.03 cm
Dimensions (L x W) 7.9 x 7.9 in. (20 x 20 cm)
Length (English) 7.9 in.
Length (Metric) 20 cm
Width (English) 7.9 in.
Width (Metric) 20 cm