Learn More
Hamilton™ HVDP 2-5 Valve for Purge and Trap Syringes
Product Code. 11510072
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11510072 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11510072 Supplier Hamilton™ Supplier No. 86789
  • ONLINE EXCLUSIVE PRICE
    197.00 EUR
    175.65 EUR / Pack of 1
    Save 21.35 EUR
    10% Off
Estimated Shipment: 14-12-2026
For use with Hamilton Purge and Trap Syringes.
  • With female luer
  • 2-5 Model
  • Aids in the analysis of drinking water samples according to EPA purge and trap concentration techniques
Specifications
Connection Type Female Luer
Color Black