missing translation for 'onlineSavingsMsg'
Learn More

Human TNF-alpha (Research Grade Adalimumab Biosimilar) Antibody, R&D Systems™
GREENER_CHOICE

Human Monoclonal Antibody

229.00 EUR - 523.00 EUR

Specifications

Antigen TNF-alpha
Clone Hu7
Dilution Neutralization 2-12 ng/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Applications Neutralization, Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18747399
GREENER_CHOICE
View Documents
R&D Systems
MAB9677-SP
25 μg
229.00 EUR
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18796818
GREENER_CHOICE
View Documents
R&D Systems
MAB9677-100
100 μg
523.00 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry.
Specifications

Specifications

TNF-alpha
Neutralization 2-12 ng/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Monoclonal
Supernatant
Human
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Human TNF-alpha
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human TNF-alpha in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Adalimumab. This product is for research use only.
Hu7
Neutralization, Flow Cytometry
Unconjugated
Human
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
7124
IgG1
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.