missing translation for 'onlineSavingsMsg'
Learn More

Human TNF-alpha (Research Grade Adalimumab Biosimilar) Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18796818
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18796818 100 μg 100µg
18747399 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18796818 Supplier R&D Systems Supplier No. MAB9677100
  • ONLINE EXCLUSIVE PRICE
    523.00 EUR
    494.55 EUR / 100µg
    Save 28.45 EUR
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Human Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Human∣Mouse∣Staphylococcus∣Streptococcus samples. It is validated for Flow Cytometry (Intracellular Staining)∣Neutralization∣Neutralization Control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF-alpha
Applications Flow Cytometry (Intracellular Staining), Neutralization, Neutralization Control
Classification Monoclonal
Clone Hu7
Conjugate Unconjugated
Dilution Neutralization 2-12 ng/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Host Species Human
Immunogen Human TNF-alpha
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity Detects human TNF-alpha in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Adalimumab. This product is for research use only.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Staphylococcus species, Streptococcus species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.