missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human T-bet/TBX21 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11637-SP
191.85 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Western Blot,Simple Western
Specifications
| T-bet/TBX21 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM | |
| E. coli -derived recombinant human T-bet, Glu326-Asp535, Accession # Q9UL17 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Simple Western | |
| 1091005 | |
| Western Blot 0.5 μg/mL, Simple Western 20 μg/mL | |
| Q9UL17 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 30009 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction