Learn More
Abcam Human RAF1 knockout HCT116 cell line
Product Code. 32141489 Shop All Abcam Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
missing translation for 'unitSize':
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
32141489 1,000,000 Cells Each
32139425 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Product Code. 32141489 missing translation for 'mfr' Abcam missing translation for 'supplierNo' ab2866191000000Cells_
 Restricted Item
Estimated Shipment: 07-12-2026
RAF1 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: select 1000000 Cells/vial.
RAF1 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells