Learn More
Invitrogen™ Human METAP1 (aa 286-359) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58202 (PA5-58202. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Tekniske data
Tekniske data
| Accession Number | P53582 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 23173 |
| Name | Human METAP1 (aa 286-359) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1700029C17Rik; AW047992; DKFZp781C0419; EC 3.4.11.18; fc84e12; im:7047238; KIAA0094; KIAA0094MAP 1; MAP 1; MAP1; MAP1A; metAP 1; Metap1; metap1 protein; MetAP1A; Methionine aminopeptidase 1; methionyl aminopeptidase 1; mKIAA0094; Peptidase M; Peptidase M 1; Putative methionine aminopeptidase 1 (MetAP 1) (Peptidase M 1); wu:fc84e12; zgc:110093 |
| Common Name | METAP1 |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.