Learn More
R&D Systems™ Human MAGEA1 (NP_004979.3) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | MAGEA1 |
| Accession Number | NP_004979.3 |
| Gene ID (Entrez) | 4100 |
| Gene Alias | Antigen MZ2-E, Cancer/testis antigen 1.1, cancer/testis antigen family 1, member 1, CT1.1, CT1.1melanoma antigen family A 1, MAGE1, MAGE-1 antigen, MAGE1A, MAGE1melanoma antigen MAGE-1, MAGE-A1, melanoma antigen family A, 1 (directs expression of antigen MZ2-E), melanoma-associated antigen 1, melanoma-associated antigen MZ2-E, MGC9326, MZ2-E |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |