missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Laminin alpha 1 N-Terminus Domain VI Antibody, R&D Systems™

Goat Polyclonal Antibody
209.00 EUR - 511.00 EUR
Specifications
| Antigen | Laminin alpha 1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725637
![]() |
R&D Systems
AF4187-SP |
25 μg |
209.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18020542
|
R&D Systems
AF4187 |
100 μg |
511.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Laminin alpha 1 Polyclonal specifically detects Laminin alpha 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry.Specifications
| Laminin alpha 1 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| LAMA, LAMA1, LAMA1 laminin, alpha 1, S-LAM-alpha | |
| LAMA1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| P25391 | |
| 284217 | |
| E. coli-derived recombinant human Laminin alpha 1 N-Terminus Domain VI Leu22-Met269 Accession # P25391 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Laminin alpha 1 N-Terminus Domain VI in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse LAMA4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title