missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Laminin alpha 1 N-Terminus Domain VI Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4187-SP
This item is not returnable.
View return policy
Description
Laminin alpha 1 Polyclonal specifically detects Laminin alpha 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry.
Specifications
| Laminin alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LAMA, LAMA1, LAMA1 laminin, alpha 1, S-LAM-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 284217 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P25391 | |
| LAMA1 | |
| E. coli-derived recombinant human Laminin alpha 1 N-Terminus Domain VI Leu22-Met269 Accession # P25391 | |
| 25 μg | |
| Primary | |
| Detects human Laminin alpha 1 N-Terminus Domain VI in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse LAMA4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction