missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IMPA1/IMP1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
184.00 EUR - 389.00 EUR
Specifications
| Antigen | IMPA1 |
|---|---|
| Clone | 984604 |
| Dilution | Immunohistochemistry 5-25μg/mL, Immunocytochemistry 8-25μg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18129420
|
R&D Systems
MAB9890-SP |
25 μg |
184.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18172731
|
R&D Systems
MAB9890-100 |
100 μg |
389.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IMPA1 Monoclonal specifically detects IMPA1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
| IMPA1 | |
| Immunohistochemistry 5-25μg/mL, Immunocytochemistry 8-25μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P29218 | |
| 3612 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 984604 | |
| Immunocytochemistry, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| EC 3.1.3.25, IMP, IMP 1, IMP1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1, MRT59, Myo-Inositol Monophosphatase 1 | |
| Synthetic peptide containing human IMPA/IMP1 Peptide Accession # P29218 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human IMPA1/IMP1 Peptide in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title