missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IMPA1/IMP1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB9890-100
This item is not returnable.
View return policy
Description
IMPA1 Monoclonal specifically detects IMPA1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| IMPA1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| EC 3.1.3.25, IMP, IMP 1, IMP1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1, MRT59, Myo-Inositol Monophosphatase 1 | |
| Synthetic peptide containing human IMPA/IMP1 Peptide Accession # P29218 | |
| 100 μg | |
| 3612 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry | |
| 984604 | |
| Immunohistochemistry 5-25μg/mL, Immunocytochemistry 8-25μg/mL | |
| P29218 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IMPA1/IMP1 Peptide in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction