Learn More
Human IL-6R alpha Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB2271R-SP
 View more versions of this product
Product Code. 18752679
  • 210.00 EUR / 25µg
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-6R alpha
Monoclonal
Unconjugated
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
Mouse
Protein A or G purified from cell culture supernatant
RUO
3570
Human
Purified
Immunocytochemistry
556704R
Immunocytochemistry 3-25 ug/mL
P08887
IL6R
E. coli-derived recombinant human IL-6 R alpha Arg387-Arg468 Accession # P08887
25 μg
Primary
Detects human IL-6 R alpha in direct ELISAs.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions