missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-6R alpha Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2271R-01M
This item is not returnable.
View return policy
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Immunocytochemistry.
Specifications
| IL-6R alpha | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3570 | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 556704R | |
| Immunocytochemistry 3-25 ug/mL | |
| P08887 | |
| IL6R | |
| E. coli-derived recombinant human IL-6 R alpha Arg387-Arg468 Accession No. P08887 | |
| 1 mg | |
| Primary | |
| Detects human IL-6 R alpha in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction