Learn More
Human IL-6R alpha Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2271R-500
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Immunocytochemistry.
Tekniske data
| IL-6R alpha | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3570 | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 556704R | |
| Immunocytochemistry 3-25 ug/mL | |
| P08887 | |
| IL6R | |
| E. coli-derived recombinant human IL-6 R alpha Arg387-Arg468 Accession No. P08887 | |
| 500 ug | |
| Primary | |
| Detects human IL-6 R alpha in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion