missing translation for 'onlineSavingsMsg'
Learn More

Human IFN-alpha I Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

199.00 EUR - 447.00 EUR

Specifications

Antigen IFN-alpha I/IFNA17
Clone 995826
Dilution Neutralization 0.6-6 ng/mL
Applications Neutralization
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18750097
GREENER_CHOICE
View Documents
R&D Systems
MAB10080-SP
25 μg
199.00 EUR
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18703527
GREENER_CHOICE
View Documents
R&D Systems
MAB10080-100
100 μg
447.00 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IFN-alpha I/IFNA17 Monoclonal specifically detects IFN-alpha I/IFNA17 in Human samples. It is validated for Neutralization.
Specifications

Specifications

IFN-alpha I/IFNA17
Neutralization 0.6-6 ng/mL
Monoclonal
Supernatant
Human
P01571
3451
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
995826
Neutralization
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
IFNA, IFNA17, IFNalpha I, IFN-alpha-17, IFN-alphaI, INFA, interferon alpha-17, Interferon alpha-88, Interferon alpha-I', Interferon alpha-T, interferon alpha-WA, interferon, alpha 17, LeIF I, LEIF2C1
Human embryonic kidney cell line HEK293-derived recombinant human IFN-alpha I, Met1-Asp189, Accession # P01571
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human IFN-alpha I in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.