missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-alpha I Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10080-SP
This item is not returnable.
View return policy
Description
IFN-alpha I/IFNA17 Monoclonal specifically detects IFN-alpha I/IFNA17 in Human samples. It is validated for Neutralization.
Specifications
| IFN-alpha I/IFNA17 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IFNA, IFNA17, IFNalpha I, IFN-alpha-17, IFN-alphaI, INFA, interferon alpha-17, Interferon alpha-88, Interferon alpha-I', Interferon alpha-T, interferon alpha-WA, interferon, alpha 17, LeIF I, LEIF2C1 | |
| Human embryonic kidney cell line HEK293-derived recombinant human IFN-alpha I, Met1-Asp189, Accession # P01571 | |
| 25 μg | |
| 3451 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 995826 | |
| Neutralization 0.6-6 ng/mL | |
| P01571 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IFN-alpha I in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction