missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-alpha/beta R1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
239.00 EUR - 622.00 EUR
Specifications
| Antigen | IFN-alpha/beta R1 |
|---|---|
| Clone | 1056928 |
| Dilution | Western Blot 2 ug/mL, Flow Cytometry 25 ug/mL |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722457
![]() |
R&D Systems
MAB2451-SP |
25 μg |
239.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18740697
![]() |
R&D Systems
MAB2451-100 |
100 μg |
622.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-alpha/beta R1 Monoclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| IFN-alpha/beta R1 | |
| Western Blot 2 ug/mL, Flow Cytometry 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| IFN-alpha/beta R1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1056928 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| AAA52730 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IFN-alpha/beta R1, Lys28-Lys436, Accession # AAA52730 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title