missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-alpha/beta R1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2451-SP
This item is not returnable.
View return policy
Description
IFN-alpha/beta R1 Monoclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| IFN-alpha/beta R1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| IFN-alpha/beta R1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IFN-alpha/beta R1, Lys28-Lys436, Accession # AAA52730 | |
| 25 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 1056928 | |
| Western Blot 2 ug/mL, Flow Cytometry 25 ug/mL | |
| AAA52730 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur