missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Frk (NP_002022.1) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | Frk |
| Accession Number | NP_002022.1 |
| Gene ID (Entrez) | 2444 |
| Gene Alias | EC 2.7.10, fyn-related kinaseProtein-tyrosine kinase 5, Gtk, GTKEC 2.7.10.2, Nuclear tyrosine protein kinase RAK, PTK5, PTK5 protein tyrosine kinase 5, Rak, RAKPTK5, tyrosine-protein kinase FRK |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction