missing translation for 'onlineSavingsMsg'
Learn More

Human FANCD2 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

221.00 EUR - 452.00 EUR

Specifications

Antigen FANCD2
Clone 1290D
Dilution Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Knockout Validated
Applications Western Blot, Immunohistochemistry, Flow Cytometry, KnockDown
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18710028
GREENER_CHOICE
View Documents
R&D Systems
MAB9369-SP
25 μg
221.00 EUR
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18723389
GREENER_CHOICE
View Documents
R&D Systems
MAB9369-100
100 μg
452.00 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FANCD2 Monoclonal specifically detects FANCD2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Knockout Validated.
Specifications

Specifications

FANCD2
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Knockout Validated
Monoclonal
Rabbit
Human
Q9BXW9
2177
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
1290D
Western Blot, Immunohistochemistry, Flow Cytometry, KnockDown
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
E. coli-derived recombinant human FANCD2 Glu11-Glu230 Accession # Q9BXW9
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human FANCD2 in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.