missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FANCD2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9369-SP
This item is not returnable.
View return policy
Description
FANCD2 Monoclonal specifically detects FANCD2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Knockout Validated.
Specifications
| FANCD2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| E. coli-derived recombinant human FANCD2 Glu11-Glu230 Accession # Q9BXW9 | |
| 25 μg | |
| Primary | |
| Detects human FANCD2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Flow Cytometry, KnockDown | |
| 1290D | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Knockout Validated | |
| Q9BXW9 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2177 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction