missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human E-Selectin/CD62E Antibody, R&D Systems™

Sheep Polyclonal Antibody
196.00 EUR - 594.00 EUR
Specifications
| Antigen | E-Selectin/CD62E |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Adhesion Blockade |
| Applications | Western Blot, Adhesion Blockade |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18735987
![]() |
R&D Systems
AF724-SP |
25 μg |
196.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18078361
|
R&D Systems
AF724 |
100 μg |
594.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E-Selectin/CD62E Polyclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for Western Blot, Adhesion Blockade.Specifications
| E-Selectin/CD62E | |
| Western Blot 0.1 ug/mL, Adhesion Blockade | |
| Polyclonal | |
| Sheep | |
| Human | |
| 6401 | |
| Chinese hamster ovary cell line CHO-derived recombinant human E-Selectin | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Selected for its ability to inhibit the block U937 cell adhesion to immobilized rhE-Selectin. In direct ELISAs, less than 5% cross-reactivity with rmE-Selectin and rrE-Selectin is observed and less than 1% cross-reactivity with rhP-Selectin and rhL-Selectin is observed. |
| LYOPH | |
| Western Blot, Adhesion Blockade | |
| Unconjugated | |
| RUO | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E | |
| SELE | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title