missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human E-Selectin/CD62E Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF724
This item is not returnable.
View return policy
Description
E-Selectin/CD62E Polyclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for Western Blot, Adhesion Blockade.
Specifications
| E-Selectin/CD62E | |
| Polyclonal | |
| Unconjugated | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6401 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Adhesion Blockade | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Adhesion Blockade | |
| SELE | |
| Chinese hamster ovary cell line CHO-derived recombinant human E-Selectin | |
| 100 μg | |
| Primary | |
| Selected for its ability to inhibit the block U937 cell adhesion to immobilized rhE-Selectin. In direct ELISAs, less than 5% cross-reactivity with rmE-Selectin and rrE-Selectin is observed and less than 1% cross-reactivity with rhP-Selectin and rhL-Selectin is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction