missing translation for 'onlineSavingsMsg'
Learn More

Human CD1e Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18786699
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18786699 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18786699 Supplier R&D Systems Supplier No. MAB7330SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD1e Monoclonal specifically detects CD1e in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD1e
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 704407
Concentration LYOPH
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P15812
Gene Alias CD1-alpha-3, CD1e, CD1e molecule, CDIEL, e polypeptide, FLJ17609, membrane-associated, R2G1, thymocyte antigen CD1E
Gene Symbols CD1E
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD1e Met1-Tyr304 Accession # P15812
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 913
Test Specificity Detects human CD1e in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CD1a, rhCD1b, rhCD1c, rhCD1d, or recombinant rat beta 2-Microglobulin is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.