missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD163 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
260.00 EUR - 473.00 EUR
Specifications
| Antigen | CD163 |
|---|---|
| Clone | 215927 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789867
![]() |
R&D Systems
FAB1607A-025 |
25 Tests |
260.00 EUR
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18788206
![]() |
R&D Systems
FAB1607A-100 |
100 Tests |
473.00 EUR
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Flow Cytometry.Specifications
| CD163 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| CD163 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CD163 in direct ELISAs and Western blots. |
| 215927 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| Q86VB7 | |
| 9332 | |
| Mouse myeloma cell line NS0-derived recombinant human CD163 Gly46-Ser1050 Accession # Q86VB7 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title