missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD163 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems FAB1607A-025
This item is not returnable.
View return policy
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD163 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9332 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 215927 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q86VB7 | |
| CD163 | |
| Mouse myeloma cell line NS0-derived recombinant human CD163 Gly46-Ser1050 Accession # Q86VB7 | |
| 25 Tests | |
| Primary | |
| Detects human CD163 in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction