Learn More
Invitrogen™ Human Caspase 6 Synthetic Peptide
Klik for at se tilgængelige muligheder
Quantity:
50 μg
Pakningsstørrelse:
50µg
Beskrivelse
Caspase-6 Synthetic Peptide, PEP-0234, from Invitrogen.
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
Tekniske data
Tekniske data
| Gene ID (Entrez) | 839 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P55212 |
| Common Name | Human Caspase 6 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion