missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbohydrate Sulfotransferase 15/CHST15 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 EUR - 402.00 EUR
Specifications
| Antigen | Carbohydrate Sulfotransferase 15/CHST15 |
|---|---|
| Clone | 388421 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778119
![]() |
R&D Systems
MAB3365-SP |
25 μg |
209.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18152031
![]() |
R&D Systems
MAB3365 |
100 μg |
402.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbohydrate Sulfotransferase 15/CHST15 Monoclonal specifically detects Carbohydrate Sulfotransferase 15/CHST15 in Human samples. It is validated for Western Blot.Specifications
| Carbohydrate Sulfotransferase 15/CHST15 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q7LFX5 | |
| 51363 | |
| Mouse myeloma cell line NS0-derived recombinant human Sulfotransferase 15/CHST15 Ser99-Thr561 Accession # Q7LFX5 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Carbohydrate Sulfotransferase 15/CHST15 in direct ELISAs and Western blots. |
| 388421 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B cell RAG associated protein (GALNAC4S-6ST), B-cell RAG-associated gene protein, BRAG, BRAGKIAA0598RP11-47G11.1, carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15, carbohydrate sulfotransferase 15, CHST15, DKFZp781H1369, EC 2.8.2.33, GalNAc4S-6ST, GALNAC4S6ST, hBRAG, MGC34346, N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase | |
| CHST15 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title