missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbohydrate Sulfotransferase 15/CHST15 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3365-SP
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 15/CHST15 Monoclonal specifically detects Carbohydrate Sulfotransferase 15/CHST15 in Human samples. It is validated for Western Blot.
Specifications
| Carbohydrate Sulfotransferase 15/CHST15 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q7LFX5 | |
| CHST15 | |
| Mouse myeloma cell line NS0-derived recombinant human Sulfotransferase 15/CHST15 Ser99-Thr561 Accession # Q7LFX5 | |
| 25 μg | |
| Primary | |
| Detects human Carbohydrate Sulfotransferase 15/CHST15 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 388421 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B cell RAG associated protein (GALNAC4S-6ST), B-cell RAG-associated gene protein, BRAG, BRAGKIAA0598RP11-47G11.1, carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15, carbohydrate sulfotransferase 15, CHST15, DKFZp781H1369, EC 2.8.2.33, GalNAc4S-6ST, GALNAC4S6ST, hBRAG, MGC34346, N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51363 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction