missing translation for 'onlineSavingsMsg'
Learn More

Human beta-3 Adrenergic R/ADRB3 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

208.00 EUR - 512.00 EUR

Specifications

Antigen beta-3 Adrenergic R/ADRB3
Clone 493324
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18735449
GREENER_CHOICE
View Documents
R&D Systems
MAB9419-SP
25 μg
208.00 EUR
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18733149
GREENER_CHOICE
View Documents
R&D Systems
MAB9419-100
100 μg
512.00 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta-3 Adrenergic R/ADRB3 Monoclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications

Specifications

beta-3 Adrenergic R/ADRB3
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
ADRB3, ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, beta3 AdrenergicR, beta-3 AdrenergicR, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960
Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3 Met1-Ser408 Accession # P13945
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Stains human beta-3 Adrenergic R/ADRB3 transfectants but not irrelevent transfectants in flow cytometry.
493324
Flow Cytometry, Immunocytochemistry
Unconjugated
Mouse
Human
P13945
155
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.