missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-3 Adrenergic R/ADRB3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9419-SP
This item is not returnable.
View return policy
Description
beta-3 Adrenergic R/ADRB3 Monoclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
| beta-3 Adrenergic R/ADRB3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADRB3, ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, beta3 AdrenergicR, beta-3 AdrenergicR, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960 | |
| Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3 Met1-Ser408 Accession # P13945 | |
| 25 μg | |
| Primary | |
| Stains human beta-3 Adrenergic R/ADRB3 transfectants but not irrelevent transfectants in flow cytometry. | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 493324 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
| P13945 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 155 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction