missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BAI1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
209.00 EUR - 501.00 EUR
Specifications
| Antigen | BAI1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Neutralization 2-8 ug/mL |
| Applications | Neutralization |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18796337
![]() |
R&D Systems
AF4969-SP |
25 μg |
209.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18068751
|
R&D Systems
AF4969 |
100 μg |
501.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAI1 Polyclonal specifically detects BAI1 in Human samples. It is validated for Neutralization.Specifications
| BAI1 | |
| Neutralization 2-8 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
| BAI1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Neutralization | |
| Unconjugated | |
| RUO | |
| O14514 | |
| 575 | |
| Chinese hamster ovary cell line CHO-derived recombinant human BAI1 Ala31-Thr879 Accession # O14514 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human BAI1 in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human BAI3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title