missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BAI1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4969
This item is not returnable.
View return policy
Description
BAI1 Polyclonal specifically detects BAI1 in Human samples. It is validated for Neutralization.
Specifications
| BAI1 | |
| Polyclonal | |
| Unconjugated | |
| O14514 | |
| BAI1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human BAI1 Ala31-Thr879 Accession # O14514 | |
| 100 μg | |
| Primary | |
| Detects human BAI1 in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human BAI3 is observed. | |
| Human | |
| IgG |
| Neutralization | |
| LYOPH | |
| Neutralization 2-8 ug/mL | |
| brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 575 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction