Learn More
Invitrogen™ Human Aminopeptidase N/ANPEP ELISA Kit
Product Code. 17185019
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17185019 5 x 96 Tests Each
17124553 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17185019 Supplier Invitrogen™ Supplier No. EH25RBX5
ELISA
Human Aminopeptidase N/ANPEP quantitates human Aminopeptidase N/ANPEP in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Aminopeptidase N/ANPEP.
CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number alanyl (membrane) aminopeptidase; Alanyl aminopeptidase; alanyl aminopeptidase, membrane; aminopeptidase; aminopeptidase M; Aminopeptidase N; aminopeptidase N/CD13; ANPEP; Apm; AP-M; Apn; AP-N; bAPN; Cd13; cluster of differentiation antigen 13 (CD13); gp150; h AP-N; hAPN; kidney aminopeptidase M; kidney Zn peptidase; KZP; Lap1; Lap-1; leucine arylaminopeptidase 1; membrane alanine aminopeptidase; membrane alanyl aminopeptidase; membrane protein p161; Microsomal aminopeptidase; myeloid plasma membrane glycoprotein CD13; P150; PEPN; rAPN; Unknown (protein for MGC:126949)
Assay Range 0.041 to 10 ng/mL
Assay Sensitivity 0.041 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 290
Gene Symbol ANPEP
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CD13
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins