Learn More
Tocris Bioscience™ HTH 01-015
GREENER_CHOICE
Product Code. p-7110850
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18778328 10 mg 10mg
18740268 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18778328 Supplier Tocris Bioscience™ Supplier No. 5622/10
Potent and selective NUAK1 inhibitor
HTH 01-015 is a potent and selective NUAK1 inhibitor (IC50 = 100 nM). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells in vitro, to a similar extent as NUAK1 knockdown models.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5,13-Dihydro-4,5,13-trimethyl-2-[[1-(4-piperidinyl)-1H-pyrazol-4-yl]amino]-6H-naphtho[2,3-e]pyrimido[5,4-b][1,4]diazepin-6-one
CAS 1613724-42-7
Quantity 10 mg
Target Other Kinase Inhibitors
Molecular Formula C26H28N8O
Formula Weight 468.55
Purity >98%