Learn More
Horst™ HSQ Model Flexible Heating Cable
Product Code. 10701943
Change view
Click to view available options
Length (Metric) Power Cord:
5m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Power Cord Unit Size
10701943 5m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10701943 Supplier Horst™ Supplier No. 21005
  • ONLINE EXCLUSIVE PRICE
    324.29 EUR
    204.95 EUR / Pack of 1
    Save 119.34 EUR
    36% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Specifications
Dimensions (L x W) 5 Lm
Dimensions (L x W x H) 5 Lm
Electrical Requirements ∼230V Voltage
Model HSQ
Length (Metric) Power Cord 5m
Product Type Flexible Heating Cable