missing translation for 'onlineSavingsMsg'
Learn More

HRAS Antibody (Y13-238) - Azide and BSA Free, Novus Biologicals™

Product Code. 18673634 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.02mg
0.20mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18673634 0.2 mg 0.20mg
18608454 0.025 mg 0.02mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18673634 Supplier Novus Biologicals Supplier No. NBP3201190.2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

HRAS Monoclonal antibody specifically detects HRAS in Human, Mouse samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRAS
Applications Immunoprecipitation
Classification Monoclonal
Clone Y13-238
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1
Host Species Rat
Immunogen Injection of rats subcutaneously with Ha-MuSV transformed NRK cells and screening of sera of individual animals to identify those with the strongest immune response against p21. Spleens were removed from the immunised animals and cell fusion of immune lymphocytes with rat myeloma cell line 210RCY3 Ag 1.2.3 (Y3) was carried out. Supernatants were harvested and assayed for antibodies to p21 by GDP binding assay or immunofluorescence on Ha-SV-transformed MDCK cells, followed by testing for the ability of the antibodies to immunoprecipitate p21.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Metastasis, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3265
Target Species Human, Mouse
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.