Learn More
Merck HPTLC Silica Gel 60, F254, Glass Plates
Specifications
Specifications
| Pore Size | 60 Å |
| Type | Glass HPTLC Plate |
| Fluorescent Indicator | F254 |
| Sorbent | Silica Gel |
| Dimensions (L x W) | 20 x 20 cm |
| Length (Metric) | 20 cm |
| Width (Metric) | 20 cm |