Learn More
SCAT HPLC Supply-Sets, V2.0
Product Code. 16413799 Shop All SCAT Europe Products
Change view
Click to view available options
Capacity (Metric):
100 mL
1 L
Color:
Brown
Clear
Material:
Glass (Round)
Glass (Square)
Glass (Youtility)
Solvent Channels:
1
2
3
Unit Size:
1 set
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Material Solvent Channels unitSize
16413799 1 L Brown Glass (Round) 1 1 set
16433789 1 L Clear Glass (Round) 1 1 set
16443789 1 L Clear Glass (Square) 1 1 set
16423799 1 L Brown Glass (Round) 2 1 set
16453789 1 L Clear Glass (Round) 2 1 set
16473789 1 L Clear Glass (Square) 2 1 set
16433799 1 L Brown Glass (Round) 3 1 set
16463789 1 L Clear Glass (Round) 3 1 set
16483789 1 L Clear Glass (Square) 3 1 set
16453799 1 L Clear Glass (Youtility) 3 1 set
17927260 100 mL Brown Glass (Round) 2 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 16413799 Supplier SCAT Supplier No. 307312
  • ONLINE EXCLUSIVE PRICE
    170.00 EUR
    146.16 EUR / 1 sets
    Save 23.84 EUR
    14% Off
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
The HPLC Supply Sets, V2.0 of the manufacturer SCAT are combinations of equipment for contamination-free supply of HPLC systems with solvents. The sets consist of a laboratory bottle with GL 45 thread, a SCAT SafetyCap as well as capillaries, suction filters and air valves.
  • Customised system with high quality set components
  • SafetyCaps of the premium manufacturer SCAT
  • Laboratory bottles
  • Price attractive in comparison with single purchase
  • Ready for use, no further components necessary
SCAT offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
Specifications
Description HPLC Supply-Set I, V2.0: 1 x Safety Cap I GL45, 1x 1000 mL Laboratory bottle round & brown glass + 1 x 1,5 m capillary 3,2 mm OD, 1 x PTFE suction filter + air valve (317010)
For Use With (Equipment) Any HPLC System
Type Complete Solvent Collection System
Solvent Channels 1
Material Glass (Round)
Includes 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle round and brown glass + 1 x 1.5 m capillary 3.2 mm OD, 1 x PTFE suction filter + air valve (317010)
Color Brown
Thread Size GL45
Capacity (Metric) 1 L