Learn More
SCAT HPLC Starter Kits - Supply, V2.0
Product Code. p-9277007 Shop All SCAT Europe Products
Change view
Click to view available options
Type:
HPLC Starter Kit 1
HPLC Starter Kit 2
HPLC Starter Kit 3
Solvent Waste Kit
Unit Size:
1 set
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
16463819 HPLC Starter Kit 1 1 set
16473819 HPLC Starter Kit 2 Pack of 4
16687493 HPLC Starter Kit 3 1 set
18656488 Solvent Waste Kit 1 set
18696458 Solvent Waste Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16463819 Supplier SCAT Supplier No. 399200
Only null left
Add to basket
Add to basket
SCAT Safety Caps are the standard for safely supplying your HPLC with solvents. They block harmful vapours and protect your HPLC system from contamination.HPLC starter kit equipped with SafetyCaps, blanking plugs, fittings and vent valves.
  • Original brand quality from the premium manufacturer SCAT
  • Initial removal equipment that can be used with all common HPLC system
  • Starter kit consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems
  • All set components can be reordered separately
starter kit, hplc system, safetycaps, blind plugs, fittings, air valve
Specifications
For Use With (Equipment) Any HPLC System
Includes 3 x SafetyCaps I (307019), 1 x SafetyCap II (307909), 1 x blind plug, 5 x fittings 1.6/2.3/3.2 mm, 4 x air valves (317010)
Thread Size GL45
For Use With (Application) Supply of HPLC Systems
Type HPLC Starter Kit 1